Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hacl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019975.3 | NP_064359.2 | 2 | 248 | ||
GenBank mRNA | AJ132139.1 | - | 2 | 246 | ||
AK005505.1 | - | 2 | 235 | |||
AK041686.1 | - | 2 | 244 | |||
AK050078.1 | - | 2 | 248 | |||
AK161488.1 | - | 228 | ||||
BC021360.1 | - | 2 | 179 | 5011451 | ||
BY340097.1 | - | 2 | 248 |