Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172436.3 | NP_766024.1 | 3 | 222 | ||
GenBank mRNA | AK019150.1 | - | 3 | 223 | ||
AK030595.1 | - | 3 | 252 | |||
AK083156.1 | - | 3 | 223 | |||
AK144958.1 | - | 3 | 256 | |||
AK147948.1 | - | 3 | 237 | |||
AK181457.1 | - | 2 | 188 | |||
AK208043.1 | - | 3 | 289 | |||
AK210791.1 | - | 2 | 188 | |||
BC013845.1 | - | 3 | 201 | |||
BC024952.1 | - | 3 | 417 | |||
BC060505.1 | - | 3 | 209 | 6311836 |