Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320445.1 | NP_001307374.1 | 3 | 340 | ||
NM_001320446.1 | NP_001307375.1 | 5 | 499 | |||
NM_015358.2 | NP_056173.1 | 4 | 413 | |||
XM_011529518.2 | XP_011527820.1 | 4 | 633 | |||
XM_017028311.1 | XP_016883800.1 | 4 | 435 | |||
XM_017028312.1 | XP_016883801.1 | 4 | 356 | |||
GenBank mRNA | AK292957.1 | - | 4 | 364 | ||
AK295133.1 | - | 3 | 332 | |||
BC007311.1 | - | 4 | 372 | |||
BC058002.1 | - | 4 | 417 | |||
BC094779.1 | - | 4 | 370 | 30527767 | ||
BC132731.1 | - | 4 | 367 | 40146753 | ||
BC144116.1 | - | 5 | 453 | |||
BU657895.1 | - | 4 | 404 | |||
D50926.2 | - | 4 | 372 |