Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304829.1 | NP_001291758.1 | 12 | 1700 | ||
NM_194292.2 | NP_919268.1 | 13 | 1780 | |||
XM_017000486.1 | XP_016855975.1 | 13 | 1546 | |||
GenBank mRNA | AK025750.1 | - | 1 | 78 | ||
AK302513.1 | - | 14 | 1530 | |||
AK307824.1 | - | 12 | 1566 | |||
AL834265.1 | - | 9 | 1043 | |||
AY359522.1 | - | 13 | 1640 | |||
BC101026.3 | - | 12 | 1493 | 40007432 | ||
BX641109.1 | - | 12 | 1691 | |||
BX648406.1 | - | 13 | 1537 |