Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021127.2 | NP_066950.1 | 2 | 309 | ||
GenBank mRNA | AK311943.1 | - | 2 | 274 | ||
BC013120.1 | - | 2 | 232 | 3858958 | ||
BC032663.1 | - | 3 | 415 | 5529754 | ||
BF667112.1 | - | 310 | ||||
D90070.1 | - | 2 | 264 | |||
KU178263.1 | - | 3 | 242 | |||
KU178264.1 | - | 2 | 91 |