Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206739.1 | NP_001193668.1 | 4 | 403 | ||
NM_014288.4 | NP_055103.3 | 3 | 286 | |||
NR_045147.1 | 3 | 286 | ||||
XM_006710514.3 | XP_006710577.3 | 3 | 530 | |||
XM_006710515.3 | XP_006710578.3 | 3 | 530 | |||
XM_017000853.1 | XP_016856342.1 | 3 | 530 | |||
XM_017000854.1 | XP_016856343.1 | 3 | 321 | |||
XM_017000855.1 | XP_016856344.1 | 2 | 299 | |||
XM_017000856.1 | XP_016856345.1 | 3 | 530 | |||
XM_017000857.1 | XP_016856346.1 | 3 | 530 | |||
GenBank mRNA | AF175306.1 | - | 3 | 177 | ||
AK309622.1 | - | 3 | 329 | |||
AK312944.1 | - | 3 | 238 | |||
BC009929.2 | - | 3 | 193 | 3505579 | ||
BC014385.1 | - | 4 | 328 | 4064625 | ||
BU853146.1 | - | 3 | 289 | |||
U37139.1 | - | 3 | 276 |