Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI27L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145249.2 | NP_660292.1 | 4 | 391 | ||
NM_206949.2 | NP_996832.1 | 4 | 345 | |||
XM_005267314.4 | XP_005267371.1 | 3 | 2936 | |||
XM_011536405.2 | XP_011534707.1 | 4 | 361 | |||
XM_011536406.1 | XP_011534708.1 | 5 | 493 | |||
XM_011536407.1 | XP_011534709.1 | 5 | 496 | |||
XM_017020961.1 | XP_016876450.1 | 4 | 242 | |||
XM_017020962.1 | XP_016876451.1 | 3 | 211 | |||
XM_017020963.1 | XP_016876452.1 | 4 | 244 | |||
GenBank mRNA | BC015423.1 | - | 4 | 355 | 4398045 | |
BU739372.1 | - | 2 | 338 |