Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKCZ SIRNA Assays ›
Gene Symbol:
PRKCZ
Gene Name:
protein kinase C zeta
Gene Aliases:
PKC-ZETA, PKC2
Chromosome Location:
Chr.1: 2050470 - 2185395 on Build GRCh38
UniGene ID:
Hs.496255
Species:
Human
siRNA ID
HSS183349
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKCZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033581.1
NP_001028753.1
4
533
NM_001033582.1
NP_001028754.1
4
430
NM_001242874.1
NP_001229803.1
4
540
NM_002744.4
NP_002735.3
7
745
XM_011541773.1
XP_011540075.1
7
741
XM_011541774.1
XP_011540076.1
4
606
XM_011541775.2
XP_011540077.1
7
741
XM_011541776.1
XP_011540078.1
4
546
XM_011541778.1
XP_011540080.1
4
459
XM_017001789.1
XP_016857278.1
7
741
XM_017001790.1
XP_016857279.1
7
741
XM_017001791.1
XP_016857280.1
4
390
XM_017001792.1
XP_016857281.1
4
546
XM_017001793.1
XP_016857282.1
4
804
XM_017001794.1
XP_016857283.1
4
459
XM_017001795.1
XP_016857284.1
4
463
XM_017001796.1
XP_016857285.1
4
546
XM_017001797.1
XP_016857286.1
2
187
XM_017001798.1
XP_016857287.1
4
459
XM_017001799.1
XP_016857288.1
4
867
XM_017001800.1
XP_016857289.1
2
187
XM_017001801.1
XP_016857290.1
2
187
XM_017001802.1
XP_016857291.1
4
546
XM_017001803.1
XP_016857292.1
7
741
XM_017001804.1
XP_016857293.1
4
456
XM_017001805.1
XP_016857294.1
4
426
GenBank mRNA
AK097627.1
-
2
166
AK294649.1
-
4
535
AK294782.1
-
4
453
AK302160.1
-
2
157
AK310372.1
-
2
187
AL542576.3
-
4
420
BC008058.2
-
7
692
2987996
BC014270.2
-
7
745
3835020
BT007082.1
-
7
584
L14283.1
-
7
584
Z15108.1
-
7
566
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
positive regulation of cell-matrix adhesion
protein phosphorylation
inflammatory response
signal transduction
transforming growth factor beta receptor signaling pathway
long-term memory
positive regulation of cell proliferation
insulin receptor signaling pathway
cell migration
peptidyl-serine phosphorylation
establishment of cell polarity
negative regulation of protein complex assembly
actin cytoskeleton reorganization
activation of phospholipase D activity
activation of protein kinase B activity
positive regulation of interleukin-4 production
negative regulation of apoptotic process
positive regulation of T-helper 2 cell differentiation
positive regulation of glucose import
negative regulation of insulin receptor signaling pathway
positive regulation of insulin receptor signaling pathway
vesicle transport along microtubule
negative regulation of peptidyl-tyrosine phosphorylation
positive regulation of NF-kappaB transcription factor activity
protein heterooligomerization
negative regulation of hydrolase activity
membrane depolarization
membrane hyperpolarization
long-term synaptic potentiation
positive regulation of ERK1 and ERK2 cascade
protein kinase C signaling
protein localization to plasma membrane
neuron projection extension
positive regulation of excitatory postsynaptic potential
positive regulation of T-helper 2 cell cytokine production
positive regulation of interleukin-5 secretion
positive regulation of interleukin-13 secretion
positive regulation of interleukin-10 secretion
protein kinase activity
protein serine/threonine kinase activity
protein kinase C activity
protein binding
ATP binding
potassium channel regulator activity
protein kinase binding
protein domain specific binding
phospholipase binding
insulin receptor substrate binding
metal ion binding
14-3-3 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0