Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRHDE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026836.1 | 2 | 842 | |||
NR_026837.1 | 1 | 1444 | ||||
GenBank mRNA | AK095156.1 | - | 1 | 461 | ||
BC040230.1 | - | 2 | 817 | |||
BC093901.1 | - | 1 | 370 | |||
BC093903.1 | - | 1 | 370 | |||
DA139014.1 | - | 1 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA348677.1 | - | 1 | 461 | Chr. 12 - 72271966 - 72272526 [-] on Build GRCh38 |