Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053670.3 | NP_446122.1 | 3, 4 | 159 | ||
GenBank mRNA | AF103950.1 | - | 1, 2 | 74 | ||
AF440799.2 | - | 3, 4 | 159 | |||
BC059117.1 | - | 3, 4 | 263 | 6891094 |
Set Membership: |
G-protein coupled receptor |