Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127192.1 | NP_001120664.1 | 2, 3 | 331 | ||
NM_001127193.1 | NP_001120665.1 | 2, 3 | 331 | |||
NM_003418.4 | NP_003409.1 | 2, 3 | 331 | |||
GenBank mRNA | AK292119.1 | - | 2, 3 | 274 | ||
AK314380.1 | - | 2, 3 | 256 | |||
BC093058.1 | - | 2, 3 | 250 | 30721134 | ||
BM786378.1 | - | 2, 3 | 318 | |||
BT019613.1 | - | 1, 2 | 125 | |||
DQ091187.1 | - | 2, 3 | 263 | |||
KU178398.1 | - | 1, 2 | 125 | |||
KU178399.1 | - | 1, 2 | 125 | |||
M28372.1 | - | 2, 3 | 227 |