Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280557.1 | NP_001267486.1 | 4 | 585 | ||
NM_017767.2 | NP_060237.2 | 11 | 2230 | |||
NM_130849.3 | NP_570901.2 | 12 | 2086 | |||
XM_006716599.1 | XP_006716662.1 | 11 | 1912 | |||
GenBank mRNA | AK000334.1 | - | 11 | 2209 | ||
AK000489.1 | - | 4 | 580 | |||
AK000695.1 | - | 11 | 2145 | |||
AK025537.1 | - | 12 | 2080 | |||
AK056900.1 | - | 3 | 671 | |||
AK290563.1 | - | 12 | 2034 | |||
BC001688.2 | - | 4 | 557 | |||
BC062625.1 | - | 12 | 2066 | 5203858 |