Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012173.3 | NP_036305.2 | 6 | 767 | ||
NM_183420.1 | NP_904356.1 | 7 | 817 | |||
NM_183421.1 | NP_904357.1 | 7 | 817 | |||
XM_011534748.2 | XP_011533050.1 | 7 | 590 | |||
XM_011534749.1 | XP_011533051.1 | 6 | 527 | |||
XM_017013313.1 | XP_016868802.1 | 7 | 641 | |||
XM_017013314.1 | XP_016868803.1 | 6 | 719 | |||
GenBank mRNA | AB070270.1 | - | 6 | 551 | ||
AF174605.1 | - | 3 | 293 | |||
AF419858.1 | - | 7 | 817 | |||
AK302334.1 | - | 7 | 576 | |||
BC020249.2 | - | 5 | 509 | 4652811 | ||
BC050393.1 | - | 7 | 670 | 5769518 |