Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135058.1 | NP_001128530.1 | 10 | 1662 | ||
NM_004086.2 | NP_004077.1 | 11 | 1338 | |||
XM_017021071.1 | XP_016876560.1 | 11 | 1572 | |||
XM_017021072.1 | XP_016876561.1 | 10 | 1711 | |||
XM_017021073.1 | XP_016876562.1 | 3 | 720 | |||
GenBank mRNA | AF006740.1 | - | 11 | 1314 | ||
AK123362.1 | - | 9 | 1467 | |||
AK130273.1 | - | 5 | 919 | |||
AK292724.1 | - | 11 | 1314 | |||
AY358900.1 | - | 10 | 1653 | |||
BC007230.1 | - | 10 | 1625 | 2824361 | ||
BC018827.2 | - | 11 | 1300 |