Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005558.3 | NP_005549.2 | 3 | 929 | ||
GenBank mRNA | AK310917.1 | - | 4 | 815 | ||
AK313388.1 | - | 3 | 712 | |||
BC009742.2 | - | 3 | 929 | 3842503 | ||
BC071890.1 | - | 3 | 990 | 6082757 | ||
BX647307.1 | - | 3 | 734 | |||
U42408.1 | - | 3 | 913 |