本ページで紹介している製品は、日本では薬機法上未承認であり、国内での販売・提供は行っておりません。
最新の製品情報はグローバルサイトをご参照ください。
The products described on this page are not approved under the Japanese regulatory framework and are not available for sale or distribution in Japan.
Please refer to the global website for the latest product information.
Celiac disease is a chronic and multiorgan autoimmune disease that affects the small bowel in genetically predisposed persons due to the ingestion of gluten.1 It can present a wide range of symptoms such as anemia, vague abdominal symptoms, esophagitis, neuropathy, ataxia, depression, short stature, osteomalacia, and multiple others.1 For the majority of patients, their celiac disease remains undiagnosed.1
Diagnostic guidelines for celiac disease recommend starting lab testing with anti-tissue transglutaminase IgA (tTG IgA) combined with the measurement of total serum IgA.1-3
For special conditions, the determination of IgA and IgG antibodies directed to deamidated gliadin peptides and anti-tissue transglutaminase IgG antibodies are recommended.1-3
Broadest portfolio for the in vitro diagnosis of wheat-related conditions. The EliA™ Celiac portfolio enables you to offer a best-in-class, workflow optimized testing algorithm to aid in the diagnosis of celiac disease:1-3
With a specificity of 98.5%, EliA Celikey IgA test is the most clinically relevant, fully automated, tissue transglutaminase IgA test—helping you help your customers to put their patients first by identifying more subjects suffering from celiac disease, correctly, the first time.4
The European Society for Paediatric Gastroenterology, Hepatology, and Nutrition (ESPGHAN) 2019 guidelines for the diagnosis of celiac disease state that if an anti-tissue transglutaminase antibody test result is greater than 10 times the manufacturers upper limit of normal range (ULN), then endoscopy can be avoided in favor of confirmatory endomysial antibody (EMA) testing.2
To demonstrate the validity of the ESPGHAN guidelines to conditionally omit biopsies, a prospective multicenter study was performed.5 In this study, it was demonstrated that with a PPV of 99% at just 2 x ULN (20 EliA U/ml) and a PPV of 100% at just 5 x ULN (50 EliA U/ml), the EliA Celikey IgA test may help reduce endoscopies more than any other automated assay. 5