ThermoFisher Scientific
Search
clear search
Recent searches Clear History
Suggestions
All Results
clear search
Search
Search Suggestions
Recent searches Clear History
お問い合わせ
炎症性腸疾患の臨床検査
医療における削減と患者転帰向上の推進
エリアの製品情報は、製品カタログをご確認ください
食事中の腹部に解剖学的イラストを重ねたイメージ
炎症性腸疾患(inflammatory bowel disease: IBD)は腸管の組織障害性の炎症を特徴とし、一般的には潰瘍性大腸炎(UC)とクローン病(CD)がこれに属します。両疾患とも、寛解期と活動期を繰り返す慢性の炎症性疾患です1。
  • IBD患者さんの約10%は過敏性腸症候群(irritable bowel syndrome: IBS)と誤診されており、またIBD患者さんの3%は正確に診断されるまでに5年以上を要します2。
  • 症状が現れてからクローン病と診断されるまでの期間は、平均9カ月です3。
  • 患者さんの予後の改善と治療の成功率向上のため、IBDの早期診断は重要です3-6。
  • エリアカルプロテクチン2のIBDの診断補助における臨床的感度および陰性的中率は、ともに100%であり、IBDとIBSの鑑別において内視鏡検査が必要かどうかの判断材料になります7。
炎症性腸疾患(IBD)に関連する検査試薬
  • エリア カルプロテクチン2 (体外診断用医薬品)
自己免疫疾患診断薬 エリアが炎症性腸疾患(IBD)の検査で選ばれる理由
大腸内視鏡検査前の予備検査として
エリアカルプロテクチン2は体外診断用医薬品として、IBDの診断補助およびUCの病態把握の補助に使用できます。
信頼できる結果
エリアカルプロテクチン2検査の感度は98~99%と高く、陰性適中率(NPV)は98.4%です。そのため、この検査は炎症性腸疾患を区別する優れた検査として使用できます8, 9。
製品カタログ
ダウンロード
参考文献
  1. Torres, J, Burisch, J, Riddle, M, Dubinsky, M et al.Preclinical disease and preventive strategies in IBD: perspectives, challenges and opportunities.Gut 2016;65(7):1061-9.
  2. Card TR, Siffledeen J, Fleming KM.Are IBD patients more likely to have a prior diagnosis of irritable bowel syndrome? Report of a case-control study in the General Practice Research Database.United European Gastroenterol J. 2014;2(6):505-12.
  3. Vavricka SR, Spigaglia SM, Rogler G, Pittet V, Michetti P, Felley C, et al.Systematic evaluation of risk factors for diagnostic delay in inflammatory bowel disease.Inflamm Bowel Dis.2012;18(3):496-505.
  4. Schoepfer AM, Dehlavi MA, Fournier N, Safroneeva E, Straumann A, Pittet V, et al.Diagnostic delay in Crohn's disease is associated with a complicated disease course and increased operation rate.Am J Gastroenterol.2013;108(11):1744-53; quiz 54.
  5. Etchevers MJ, Aceituno M, Sans M. Are we giving azathioprine too late? The case for early immunomodulation in inflammatory bowel disease.World J Gastroenterol.2008;14(36):5512-8.
  6. Markowitz J. Early inflammatory bowel disease: different treatment response to specific or all medications? Dig Dis.2009;27(3):358-65.
  7. 松岡克善ら. 医学と薬学 2007; 74(6): 717-726
  8. Wulandari A A, Schulz J, Hartmann F, Dignass A, Stein J. Poster No 119 presented at 11th Congress of ECCO, Amsterdam, Netherlands, 2016 e 48. 
  9. Thermo Fisher Scientific.Internal Study.