Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342508.1 | NP_001329354.1 | 6 - 7 | 2104 | 72 | |
NM_120021.2 | NP_195572.2 | 6 - 7 | 1656 | 72 | ||
NM_179236.3 | NP_849567.2 | 6 - 7 | 2104 | 72 | ||
GenBank mRNA | AY265959.1 | - | 6 - 7 | 1696 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |