Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343274.1 | NP_001318555.1 | 5 - 6 | 779 | 91 | |
NM_001343275.1 | NP_001331012.1 | 5 - 6 | 710 | 91 | ||
NM_001343276.1 | NP_001331011.1 | 5 - 6 | 710 | 91 | ||
NM_121351.5 | NP_196852.3 | 4 - 5 | 746 | 91 | ||
GenBank mRNA | BT012225.1 | - | 4 - 5 | 473 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |