Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340873.1 | NP_001328391.1 | 3 - 4 | 851 | 89 | |
NM_117441.4 | NP_193103.2 | 3 - 4 | 851 | 89 | ||
GenBank mRNA | BT029541.1 | - | 3 - 4 | 786 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |