Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203666.1 | NP_001190595.1 | 16 - 17 | 1528 | 78 | |
NM_125617.3 | NP_201029.1 | 18 - 19 | 1665 | 78 | ||
GenBank mRNA | FJ708810.1 | - | 18 - 19 | 1368 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |