Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113032.4 | NP_188774.2 | 6 - 7 | 628 | 77 | |
GenBank mRNA | AY064046.1 | - | 6 - 7 | 603 | 77 | |
AY096383.1 | - | 6 - 7 | 530 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |