Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332726.1 | NP_001319086.1 | 5 - 6 | 923 | 100 | |
NM_102433.1 | NP_564257.1 | 4 - 5 | 614 | 100 | ||
GenBank mRNA | AF369575.1 | - | 4 - 5 | 614 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |