Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014856.1 | NP_001014856.1 | 3 - 4 | 322 | 152 | |
GenBank mRNA | BC134466.1 | - | 3 - 4 | 345 | 152 | |
BT021024.1 | - | 3 - 4 | 322 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |