Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bli-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026369.6 | NP_001021540.1 | 7 - 8 | 1274 | 72 | |
NM_001026372.7 | NP_001021543.1 | 7 - 8 | 1280 | 72 | ||
NM_001316849.3 | NP_001303778.1 | 7 - 8 | 1280 | 72 | ||
NM_001316850.3 | NP_001303779.1 | 7 - 8 | 1276 | 72 | ||
NM_001316852.3 | NP_001303781.1 | 7 - 8 | 1276 | 72 | ||
NM_001372959.2 | NP_001360008.1 | 7 - 8 | 1281 | 72 | ||
NM_001380517.2 | NP_001367786.1 | 7 - 8 | 1262 | 72 | ||
NM_001380518.2 | NP_001367785.1 | 8 - 9 | 1324 | 72 | ||
NM_001380521.1 | NP_001368583.1 | 6 - 7 | 1116 | 72 | ||
NM_001380523.1 | NP_001367787.1 | 6 - 7 | 1116 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |