Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069744.7 | NP_502145.2 | 6 - 7 | 2514 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |