Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043526.1 | XP_017899015.1 | 3 - 4 | 292 | 98 | |
GenBank mRNA | JO528756.2 | - | 2 - 3 | 281 | 98 | |
KA325906.1 | - | 2 - 3 | 445 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |