Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079705.3 | NP_524429.1 | 1 - 2 | 603 | 71 | |
GenBank mRNA | AF181656.1 | - | 1 - 2 | 555 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |