Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260085.2 | NP_001247014.1 | 18 - 19 | 13802 | 68 | |
NM_001260086.3 | NP_001247015.1 | 19 - 20 | 13883 | 68 | ||
NM_001260087.3 | NP_001247016.2 | 19 - 20 | 14123 | 68 | ||
NM_001316573.1 | NP_001303502.1 | 17 - 18 | 13748 | 68 | ||
NM_001316574.1 | NP_001303503.1 | 16 - 17 | 8654 | 68 | ||
NM_176443.2 | NP_788620.1 | 15 - 16 | 7883 | 68 | ||
NM_176444.2 | NP_788621.1 | 17 - 18 | 8687 | 68 | ||
NM_176445.2 | NP_788622.1 | 16 - 17 | 13728 | 68 | ||
NM_176446.2 | NP_788623.1 | 15 - 16 | 13684 | 68 | ||
NM_176447.2 | NP_788624.1 | 19 - 20 | 13850 | 68 | ||
NM_176448.2 | NP_788625.1 | 16 - 17 | 13624 | 68 | ||
NM_176449.3 | NP_788626.2 | 15 - 16 | 13568 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |