Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkchb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044806.1 | NP_001038271.1 | 12 - 13 | 1738 | 83 | |
XM_005160500.4 | XP_005160557.1 | 12 - 13 | 1963 | 83 | ||
GenBank mRNA | BC163148.1 | - | 12 - 13 | 1836 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |