Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zic2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001820.1 | NP_001001820.1 | 2 - 3 | 1367 | 65 | |
GenBank mRNA | AF207751.2 | - | 2 - 3 | 1367 | 65 | |
BC098556.1 | - | 2 - 3 | 1380 | 65 | ||
BC171625.1 | - | 2 - 3 | 1363 | 65 | ||
BC171629.1 | - | 2 - 3 | 1363 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |