Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000381887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000381887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000381887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000381887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000383917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
ENST00000495872.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03038240.hg.1 |
TC03002159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03038240.hg.1 |
ENST00000296328.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03038240.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03038240.hg.1 |
ENST00000535858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03038240.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000381887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000381887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000381887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000381887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000383917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000413584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000493566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
ENST00000495872.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013701.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
ENST00000296328.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
ENST00000428095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
ENST00000429160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
NM_015562.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202405.hg.1 |
TC0300013701.hg.1 |
Clariom D TC |