Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATP11A upstream neighbor | ATP11AUN |
C13orf35, SMABLO1 |
-53.1kb | Anti-sense | Chr. 13 - 112647044 - 112684497 on NCBI Build GRCh38 |
Downstream | ATP11A antisense RNA 1 | ATP11A-AS1 | 7.4kb | Sense | Chr. 13 - 112745449 - 112754731 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |