Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200008871.mm.1 |
NM_010469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008871.mm.1 |
NM_010469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008871.mm.1 |
TC0200001097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008872.mm.1 |
NM_010469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008872.mm.1 |
NM_010469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008872.mm.1 |
TC0200001097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008869.mm.1 |
NM_010469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008869.mm.1 |
NM_010469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008869.mm.1 |
TC0200001097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008873.mm.1 |
NM_010469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008873.mm.1 |
NM_010469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008873.mm.1 |
TC0200001097.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008874.mm.1 |
NM_010469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008874.mm.1 |
NM_010469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008874.mm.1 |
TC0200001097.mm.1 |
MTA TC |