Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026842.4 | NP_081118.4 | 7 - 8 | 1451 | 77 | |
GenBank mRNA | AK049298.1 | - | 7 - 8 | 1206 | 77 | |
AK187729.1 | - | 1 - 2 | 65 | 77 | ||
AK198908.1 | - | 1 - 2 | 65 | 77 | ||
AK209853.1 | - | 1 - 2 | 65 | 77 | ||
BC010213.1 | - | 4 - 5 | 794 | 77 | ||
BC026847.1 | - | 7 - 8 | 1434 | 4015641 | 77 | |
BC027375.1 | - | 1 - 2 | 154 | 77 | ||
BC051098.1 | - | 6 - 7 | 1079 | 77 | ||
BC080667.1 | - | 7 - 8 | 1412 | 4935292 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |