Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111110.2 | NP_001104580.1 | 5 - 6 | 777 | 69 | |
NM_001284519.1 | NP_001271448.1 | 6 - 7 | 939 | 69 | ||
NM_001284520.1 | NP_001271449.1 | 4 - 5 | 432 | 69 | ||
NM_007717.5 | NP_031743.3 | 4 - 5 | 432 | 69 | ||
GenBank mRNA | AB061276.1 | - | 5 - 6 | 761 | 69 | |
AB061277.1 | - | 6 - 7 | 923 | 69 | ||
AK040274.1 | - | 3 - 4 | 382 | 69 | ||
AK083394.1 | - | 3 - 4 | 466 | 69 | ||
BC028474.1 | - | 5 - 6 | 787 | 69 | ||
BC055079.1 | - | 3 - 4 | 429 | 30077348 | 69 | |
CA577904.1 | - | 3 - 4 | 417 | 69 | ||
D21826.1 | - | 3 - 4 | 404 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |