Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017370.2 | NP_059066.1 | 2 - 3 | 243 | 81 | |
GenBank mRNA | AK150430.1 | - | 2 - 3 | 136 | 81 | |
AK150830.1 | - | 2 - 3 | 136 | 81 | ||
AK152295.1 | - | 2 - 3 | 136 | 81 | ||
AK152698.1 | - | 2 - 3 | 136 | 81 | ||
AK159676.1 | - | 2 - 3 | 135 | 81 | ||
BC138872.1 | - | 2 - 3 | 118 | 8861894 | 81 | |
M96827.1 | - | 2 - 3 | 133 | 81 | ||
S67972.1 | - | 2 - 3 | 134 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |