Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164222.1 | NP_001157694.1 | 4 - 5 | 888 | 68 | |
NM_023420.2 | NP_075909.1 | 4 - 5 | 888 | 68 | ||
GenBank mRNA | AF232932.1 | - | 4 - 5 | 902 | 68 | |
AF232934.1 | - | 4 - 5 | 902 | 68 | ||
AK012989.1 | - | 4 - 5 | 869 | 68 | ||
AK030305.1 | - | 5 - 6 | 796 | 68 | ||
AK038061.1 | - | 4 - 5 | 878 | 68 | ||
AK038803.1 | - | 4 - 5 | 869 | 68 | ||
AK047100.1 | - | 4 - 5 | 885 | 68 | ||
AK080958.1 | - | 4 - 5 | 886 | 68 | ||
AK163624.1 | - | 4 - 5 | 878 | 68 | ||
BC016197.1 | - | 4 - 5 | 851 | 4502118 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |