Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026813.1 | NP_081089.1 | 3 - 4 | 507 | 118 | |
NM_027805.2 | NP_082081.1 | 3 - 4 | 507 | 118 | ||
GenBank mRNA | AK018403.1 | - | 3 - 4 | 427 | 118 | |
AK083335.1 | - | 3 - 4 | 499 | 118 | ||
AK084684.1 | - | 3 - 4 | 508 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |