Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026666.3 | NP_080942.1 | 12 - 13 | 2425 | 146 | |
GenBank mRNA | AK147571.1 | - | 12 - 13 | 2426 | 146 | |
BC060723.1 | - | 10 - 11 | 1353 | 146 | ||
BC098372.1 | - | 10 - 11 | 1585 | 30363389 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |