Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028451.1 | NP_082727.1 | 14 - 15 | 2609 | 59 | |
GenBank mRNA | AK129202.1 | - | 3 - 4 | 357 | 59 | |
BC063078.1 | - | 1 - 2 | 44 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |