Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | X98112.1 | - | 2 - 3 | 614 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |