Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_001007465.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_001007465.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_001164569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_001164569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_001164570.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_001164570.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_001164571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_001164571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_026097.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
NM_026097.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030910.mm.1 |
TC1100003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_001007465.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_001007465.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_001164569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_001164569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_001164570.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_001164570.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_001164571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_001164571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_026097.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
NM_026097.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030909.mm.1 |
TC1100003369.mm.1 |
MTA TC |