Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139445.2 | NP_001132917.1 | 1 - 2 | 159 | 116 | |
GenBank mRNA | FJ238618.1 | - | 1 - 2 | 160 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |