Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LLGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098677.2 | XP_014954163.2 | 4 - 5 | 439 | 79 | |
XM_027974801.1 | XP_027830602.1 | 4 - 5 | 430 | 79 | ||
XM_027974802.1 | XP_027830603.1 | 4 - 5 | 430 | 79 | ||
XM_027974803.1 | XP_027830604.1 | 4 - 5 | 430 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |