Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170977.1 | NP_001164448.1 | 2 - 3 | 360 | 68 | |
XM_008264012.2 | XP_008262234.1 | 2 - 3 | 383 | 68 | ||
XM_017345508.1 | XP_017200997.1 | 2 - 3 | 390 | 68 | ||
GenBank mRNA | M28655.1 | - | 2 - 3 | 352 | 68 | |
X57209.1 | - | 360 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |