Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106059.1 | NP_001099529.1 | 4 - 5 | 827 | 85 | |
GenBank mRNA | BC168852.1 | - | 4 - 5 | 810 | 7443262 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |