Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246362.2 | XP_006246424.1 | 1 - 2 | 485 | 67 | |
XM_006246363.2 | XP_006246425.1 | 1 - 2 | 495 | 67 | ||
XM_006246364.2 | XP_006246426.1 | 1 - 2 | 489 | 67 | ||
XM_006246365.2 | XP_006246427.1 | 1 - 2 | 485 | 67 | ||
XM_008767741.1 | XP_008765963.1 | 1 - 2 | 2309 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |