Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199269.1 | NP_954890.1 | 10 - 11 | 2208 | 58 | |
XM_008764688.1 | XP_008762910.1 | 9 - 10 | 2997 | 58 | ||
GenBank mRNA | AY371924.1 | - | 10 - 11 | 2208 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |