Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181380.2 | NP_852045.1 | 1 - 2 | 48 | 113 | |
GenBank mRNA | AF532860.1 | - | 1 - 2 | 48 | 113 | |
AX751537.1 | - | 1 - 2 | 30 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |